PHL82257
trans-Aconitic acid
phyproof® Reference Substance
Synonym(s):
trans-Propene-1,2,3-tricarboxylic acid
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (HPLC)
manufacturer/tradename
PhytoLab
mp
190 °C (dec.) (lit.)
application(s)
food and beverages
format
neat
SMILES string
OC(=O)C\C(=C/C(O)=O)C(O)=O
InChI
1S/C6H6O6/c7-4(8)1-3(6(11)12)2-5(9)10/h1H,2H2,(H,7,8)(H,9,10)(H,11,12)/b3-1+
InChI key
GTZCVFVGUGFEME-HNQUOIGGSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Biochem/physiol Actions
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service