Skip to Content
Merck
All Photos(4)

Key Documents

P9875

Sigma-Aldrich

Phenolphthalein bisphosphate tetrasodium salt

≥92.5% purity, powder

Synonym(s):

Phenolphthalein diphosphate tetrasodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$65.40
5 G
$179.00
25 G
$627.00

$65.40


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$65.40
5 G
$179.00
25 G
$627.00

About This Item

Linear Formula:
C20H12O10P2Na4
CAS Number:
Molecular Weight:
566.21
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$65.40


Estimated to ship on26 May 2025


Request a Bulk Order

Product Name

Phenolphthalein bisphosphate tetrasodium salt, ~95%

Assay

~95%

Quality Level

form

powder

impurities

Phenolphthalein, none detected
5-10% solvent (actual content on label)

solubility

water: 50 mg/mL, clear, colorless to faint yellow or tan

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)Oc1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc(OP([O-])([O-])=O)cc4

InChI

1S/C20H16O10P2.4Na/c21-19-17-3-1-2-4-18(17)20(28-19,13-5-9-15(10-6-13)29-31(22,23)24)14-7-11-16(12-8-14)30-32(25,26)27;;;;/h1-12H,(H2,22,23,24)(H2,25,26,27);;;;/q;4*+1/p-4

InChI key

HTMKLCRJVFFVDW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Phenolphthalein bisphosphate tetrasodium salt has been used as a component of modified Baird Parker (BP) agar to determine the expression of phosphatases of Staphylococcus aureus ATCC12600.[1] It has also been used as a component of modified Tryptose phosphate/phenolphthalein/methyl green (M-TPMG) agar to detect enteroinvasive Escherichia coli (EIEC) apyrase activity.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ala160 and His116 residues are involved in activity and specificity of apyrase, an ATP-hydrolysing enzyme produced by enteroinvasive Escherichia coli
Serena Sarli
Microbiology (2005)
Phosphorylation controls the functioning of Staphylococcus aureus isocitrate dehydrogenase ? favours biofilm formation
U. Venkateswara Prasad
Journal of Enzyme Inhibition, 655-661 (2015)
U Venkateswara Prasad et al.
Journal of enzyme inhibition and medicinal chemistry, 30(4), 655-661 (2015-03-07)
Isocitrate dehydrogenase (IDH) gene from Staphylococcus aureus ATCC12600 was cloned, sequenced and characterized (HM067707). PknB site was observed in the active site of IDH; thus, it was predicted as IDH may be regulated by phosphorylation. Therefore, in this study, PknB

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service