Skip to Content
Merck
All Photos(1)

Key Documents

O3878

Supelco

Oxeladin citrate salt

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H33NO3 · C6H8O7
CAS Number:
Molecular Weight:
527.60
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)CCOCCOC(=O)C(CC)(CC)c1ccccc1

InChI

1S/C20H33NO3.C6H8O7/c1-5-20(6-2,18-12-10-9-11-13-18)19(22)24-17-16-23-15-14-21(7-3)8-4;7-3(8)1-6(13,5(11)12)2-4(9)10/h9-13H,5-8,14-17H2,1-4H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KVKJFNUGVOFNGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alaa El-Gindy
Farmaco (Societa chimica italiana : 1989), 60(8), 689-699 (2005-07-19)
Two high performance liquid chromatographic (HPLC) methods are presented for the determination of oxeladin citrate (OL) and oxybutynin hydrochloride (OB) and their degradation products. The first method was based on HPLC separation of OL from its degradation product using a
Determination of oxeladin in human sera by gas--liquid chromatography with thermionic detection.
B Salvesen et al.
Journal of chromatography, 225(2), 463-468 (1981-10-09)
[Determination of oxeladin citrate in drug combinations].
C Knopp et al.
Die Pharmazie, 37(1), 76-77 (1982-01-01)
C Lartigue-Mattei et al.
Biomedical chromatography : BMC, 9(1), 32-35 (1995-01-01)
A capillary gas chromatographic method with mass-selective detection was developed for the determination of oxeladin in human plasma. Plasma samples (1 mL) were alkalinized and extracted using 5mL of hexane: isoamyl alcohol (99:1). The method was demonstrated to be sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service