Skip to Content
Merck
All Photos(1)

Key Documents

C9781

Sigma-Aldrich

Tyr-C-Peptide human

≥95% (HPLC)

Synonym(s):

Tyr-Arg-Arg-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-Lys-Arg, Tyr-Insulin C chain

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
$334.00

$334.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.1 MG
$334.00

About This Item

Empirical Formula (Hill Notation):
C162H268N50O54
CAS Number:
Molecular Weight:
3780.16
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

$334.00


Check Cart for Availability

Request a Bulk Order

Product Name

Tyr-C-Peptide human, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder

composition

Peptide content, ~65%

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... INS(3630)

Amino Acid Sequence

Tyr-Arg-Arg-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-Lys-Arg

Biochem/physiol Actions

Tyr-C-Peptide is derived from genetically modified Arg32Tyr human pro-insulin mutants by trypsin and carboxypeptidase B codigestion.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Sun et al.
Applied biochemistry and biotechnology, 55(3), 167-174 (1995-12-01)
C-peptide radioimmunoassay (C-peptide RIA) is widely used in determination of pancreatic B-cell secretion activity. 125I labeled Tyr-C-peptide is indispensable in C-peptide RIA kit. Herein we discuss a way of obtaining recombinant Tyr-C-peptide. Arg32Tyr human pro-insulin mutant (R32Y-proinsulin) gene was constructed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service