Skip to Content
Merck
All Photos(1)

Documents

O0225800

Oxfendazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methyl 5-(phenylsulfinyl)-benzimidazol-2-carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O3S
CAS Number:
Molecular Weight:
315.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxfendazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)Nc1nc2cc(ccc2[nH]1)S(=O)c3ccccc3

InChI

1S/C15H13N3O3S/c1-21-15(19)18-14-16-12-8-7-11(9-13(12)17-14)22(20)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)

InChI key

BEZZFPOZAYTVHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxfendazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D N Ali et al.
International journal for parasitology, 23(4), 477-484 (1993-07-01)
Within 3 h of intra-ruminal (IR) administration oxfendazole (OFZ) was extensively associated with digesta particulate material. This association appeared to be a passive adsorption/desorption since sheep given dose equivalent OFZ which was contained solely in rumen particulate digesta (obtained from
Xiao-Lin Hou et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 962, 20-29 (2014-06-03)
A simple, sensitive and reliable analytical method was developed for the simultaneous determination of 38 veterinary drugs (18 sulfonamides, 11 quinolones and 9 benzimidazoles) and 8 metabolites of benzimidazoles in bovine milk by ultra high performance liquid chromatography-positive electrospray ionization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service