Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8983

Silicon nitride

NIST® RM 8983

Sign Into View Organizational & Contract Pricing

Select a Size

4.5 G
$1,650.00

$1,650.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
4.5 G
$1,650.00

About This Item

Linear Formula:
Si3N4
CAS Number:
Molecular Weight:
140.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,650.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

manufacturer/tradename

NIST®

density

3.44 g/mL at 25 °C (lit.)

application(s)

glass & ceramic

format

matrix material

SMILES string

N12[Si]34N5[Si]16N3[Si]25N46

InChI

1S/N4Si3/c1-5-2-6(1)3(5)7(1,2)4(5)6

InChI key

HQVNEWCFYHHQES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of RM 8983 comprises a bottle containing approximately 4.5 g of silicon nitride powder.

Application

The Reference Material silicon nitride powder is intended for use as an analytical standard for determining nitrogen, oxygen, and carbon in silicon nitride powder using the high-temperature combustion method.

Features and Benefits

  • The non-certified values for total nitrogen, oxygen, and carbon are reported on an as-received basis and expressed as mass fraction.
  • The period of validity, maintenance of non-certified values, and storage instructions are provided on NIST certificate.

Other Notes

Reference/Informational value is provided for the following analytes.
Nitrogen (N), Oxygen (0), Carbon (C)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service