Skip to Content
Merck
All Photos(2)

Key Documents

NIST1900

Silicon nitride powder - specific surface area standard

NIST® SRM® 1900

Synonym(s):

Silicon nitride

Sign Into View Organizational & Contract Pricing

Select a Size

4 G
$1,080.00

$1,080.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
4 G
$1,080.00

About This Item

Linear Formula:
Si3N4
CAS Number:
Molecular Weight:
140.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,080.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

powder

packaging

pkg of 4 g

manufacturer/tradename

NIST®

density

3.44 g/mL at 25 °C (lit.)

application(s)

glass & ceramic

SMILES string

N12[Si]34N5[Si]16N3[Si]25N46

InChI

1S/N4Si3/c1-5-2-6(1)3(5)7(1,2)4(5)6

InChI key

HQVNEWCFYHHQES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 1900 consists of a bottle containing ~ 4g of silicon nitride powder.

SRM 1900_cert

SRM 1900_SDS

Application

Silicon nitride powder - specific surface area standard reference material (SRM) is intended to calibrate instruments used to measure the specific surface area of materials, falling within a range of 0.1_m2/g to 1000_m2/g.

Features and Benefits

  • SRM 1900 is available with certified specific surface area.  
  • Certified value is measured using a static volumetric type Brunauer–Emmett–Teller (BET) (nitrogen gas) instrument using a pellet cell. 
  • The NIST certificate is provided with storage, handling, and use instructions

Other Notes

Reference/Informational value is provided for the following analytes.
Nitrogen (N), Oxygen (O2), Water (H2O), Total hydrocarbon

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service