Skip to Content
Merck
All Photos(2)

Key Documents

NIST682

High-purity zinc

NIST® SRM® 682

Synonym(s):

Zinc

Sign Into View Organizational & Contract Pricing

Select a Size

1 BLOCK
$1,390.00

$1,390.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 BLOCK
$1,390.00

About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,390.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

vapor pressure

1 mmHg ( 487 °C)

packaging

pkg of block

manufacturer/tradename

NIST®

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

High-purity zinc is a semicircular bar segment with a 57 mm (2 1/4 inch) diameter, 25.4 mm (1 inch) depth at the mid-diameter, and 19 mm (3/4 inch) length. For more information, please refer to the COA and SDS.

SRM 682_cert

SRM 682_SDS

Application

Standard Reference Material(SRM) is used to calibrate instruments and evaluate chemical methods used inzinc analysis.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Beryllium (Be), Bismuth (Bi), Boron (B), Cadmium (Ca), Copper (Cu), Iron (Fe), Lead (Pb), Silver (Ag), Thallium (Tl), Tin (Sn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service