Skip to Content
Merck
All Photos(1)

Key Documents

NIST363

Low alloy steel

NIST® SRM® 363, Cr-V (mod.)

Sign Into View Organizational & Contract Pricing

Select a Size

150 G
$1,420.00

$1,420.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
150 G
$1,420.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,420.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 150 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in chemical methods of analysis. SRM 363 isin the form of chips sized between 0.5 mm and 1.18 mm sieve openings (35 mesh and 16 mesh)

SRM 363_cert

SRM 363_SDS

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Molybdenum (Mo), Antimony (Sb), Neodymium (Nd), Arsenic (As), Nickel (Ni), Calcium (Ca), Niobium (Nb), Carbon (C), Phosphorus (P), Cerium (Ce), Silicon (Si), Chromium (Cr), Silver (Ag), Cobalt (Co), Sulfur (S), Copper (Cu), Tin (Sn), Gold (Au), Titanium (Ti), Lead (Pb), Tungsten (W), Magnesium (Mg), Vanadium (V), Manganese (Mn), Zirconium (Zr), Boron (B)

Reference/Informational value is provided for the following analytes.
Bismuth (Bi), Oxygen (O), Germanium (Ge), Praseodymium (Pr), Hafnium (Hf), Selenium (Se), Hydrogen (H), Strontium (Sr), Iron (Fe), Tantalum (Ta), Lanthanum (La), Tellurium (Te), Nitrogen (N), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service