Select a Size
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 vials x 4 g ea
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SRM 1887B_cert
SRM 1887B_SDS
Application
Features and Benefits
- Accompanied with certificate documenting NIST- certified mass fraction values, and coverage factor for the constituents in portland cement.
- The reference and information values of constituents are reported as mass fraction.
- The certified value is metrologically traceable to the SI unit of mass fraction, expressed as a percentage.
- The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.
Other Notes
Silicon dioxide (SiO2), Aluminium oxide (Al2O3), Ferric oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sulphur Trioxide (SO3), Sodium oxide (Na2O), Potassium oxide (K2O), Titanium dioxide (TiO2), Phosphorus pentoxide (P2O5), Zinc oxide (ZnO), Manganese(III) oxide (Mn2O3), Chlorine (Cl), Chromium(III) oxide (Cr2O3), Strontium oxide (SrO)
Reference/Informational value is provided for the following analytes.
Sulfide sulfur (S2-), Free Calcium oxide (CaO), Fluoride (F–), Barium oxide (BaO)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service