Skip to Content
Merck
All Photos(1)

Key Documents

NIST3384

Ground Asian Ginseng (Panax Ginseng C.A. Meyer) Rhizome

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$1,210.00

$1,210.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
15 MG
$1,210.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,210.00


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 15 mg

manufacturer/tradename

NIST®

application(s)

agriculture

format

matrix material

General description

One unit of SRM 3384 contains five packets, with each measuring ~ 3 grams of ground Asian ginseng rhizome.
For more information, please refer to the SDS and COA.
SRM 3384_cert
SRM 3384_SDS

Application
SRM is intended for evaluating analytical methods used to determine ginsenosides and trace elements in ground Asian ginseng (Panax ginseng C.A. Meyer) rhizomes, as well as in similar matrices.
Also, it can be employed for quality assurance when determining values for in-house reference materials.

Features and Benefits

  • NISTcertified mass fraction values for each analyte in SRM 3384 are provided on adry-mass basis and values are metrologically traceable to the SI unit of mass,expressed as milligrams per kilogram (mg/kg).
  • Non-Certified mass fraction values for analytes in SRM 3384 areprovided on a dry-mass basis and values are metrologically traceable to SI unitof mass fraction expressed as milligrams per gram or milligrams per kilogram.
  • Period of validity, source, instructions for storage, handling,and uses are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Arsenic (As), Lead (Pb)

Reference/Informational value is provided for the following analytes.
Ginsenoside Rb1 (C54H92O23), Ginsenoside Re (C48H82O18), Ginsenoside Rb2 (C53H90O22), Ginsenoside Rf (C42H72O14), Ginsenoside Rc (C53H90O22), Ginsenoside Rg1 (C42H72O14), Ginsenoside Rd (C48H82O18), Cadmium (Cd)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service