Skip to Content
Merck
All Photos(1)

Key Documents

NIST3385

Asian Ginseng (Panax ginseng) Extract

NIST®SRM®

Synonym(s):

Kudzu rhizome

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,000.00

$1,000.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$1,000.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,000.00


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 5 g

manufacturer/tradename

NIST®

application(s)

food and beverages

format

matrix material

General description

One unit of SRM 3385 contains five packets, each measuring ~ 1 gram of ground Asian ginseng extract.
For more information, please refer to the SDS and COA.
SRM 3385_cert
SRM 3385_SDS

Application
Asian Ginseng SRM is intended for evaluating analytical methods employed in determining ginsenosides and elements in ground Asian ginseng extract and other materials of a similar matrix.
Also, it can be used for quality assurance when determining values for in-house reference materials.

Features and Benefits

  • Certified Mass Fraction values for ginsenosides are provided on anas-received basis. The values are metrologically traceable to the SI unit forchemical mass fraction and expressed as milligrams per gram.
  • Non-Certified Mass Fraction Values forGinsenoside Rc and elements are provided.
  • NISTcertificate is provided with expiration details, instructions for storage,handling, and use.

Other Notes

Certified for the analytes listed below.
Ginsenoside Rb1, Ginsenoside Re, Ginsenoside Rb2, Ginsenoside Rf, Ginsenoside Rd, Ginsenoside Rg1

Reference/Informational value is provided for the following analytes.
Arsenic (As), Ginsenoside Rc, Cadmium (Cd), Lead (Pb)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service