Skip to Content
Merck
All Photos(1)

Key Documents

NIST2579A

Lead paint films for Building surfaces (SRM 2570 through 2575)

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

150 G
$3,270.00

$3,270.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
150 G
$3,270.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$3,270.00


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 150 g

manufacturer/tradename

NIST®

application(s)

coatings

format

matrix material

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service