NIST2571
Lead paint film for portable x-ray fluorescence analyzers
NIST® SRM® 2571, nominal 3.5 mg/cm2 (color code: yellow)
Select a Size
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 1 +blank
manufacturer/tradename
NIST®
technique(s)
XRF: suitable
application(s)
coatings
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
SRM 2571_cert
SRM 2571_SDS
Other Notes
Lead (Pb)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service