Skip to Content
Merck
All Photos(1)

Key Documents

NIST2492

Bingham paste mixture for rheological measurements

NIST® SRM® 2492

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,270.00

$1,270.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$1,270.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,270.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of kit for two batches

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in calibrating rheometers for measuring the rheological properties of cement and concrete. A unit of SRM 2492 consists of three containers, one glass bottle of corn syrup (500 g) and two plastic jars of limestone powder (600 g each). For more information, please refer to the SDS and COA.

SRM 2492_cert

SRM 2492_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service