Skip to Content
Merck
All Photos(1)

Key Documents

MAK457

Sigma-Aldrich

Caspase-3 Assay Kit

Sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$582.00

$582.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$582.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$582.00


Check Cart for Availability

Request a Bulk Order

Quality Level

input

cell lysate
tissue

detection method

fluorometric

relevant disease(s)

neurological disorders; cancer

storage temp.

−20°C

General description

CPP32 is a gene encoding a member of the caspase family, crucial for both the initiation and execution phases of apoptosis, a fundamental process in various physiological and pathological events, ranging from normal fetal development to diseases such as cancer, organ failure, and neurodegenerative diseases. Human caspases are categorized into three functional groups: cytokine activation, apoptosis initiation, and apoptosis execution. Among these, Caspase 3, also known as CPP32, Apopain, Yama, and PARP cleavage protease, is extensively studied and regulates numerous cellular events. Its activity is modulated by various factors, including APAF1, CFLAR/FLIP, NOL3/ARC, and members of the inhibitor of apoptosis (IAP) family. Caspase 3 undergoes self-proteolysis and/or cleavage by other proteases, resulting in its activation. It subsequently cleaves and activates other caspases and targets in cells. Variations in Caspase 3 levels are observed in cells with different lifespans and in diseases like Alzheimer′s. Caspase-3 serves as a vital biomarker in apoptosis assessment and aids in understanding its underlying mechanisms.

Application

The Caspase-3 Assay Kit may be used for:
  • Apoptosis Inducer and Inhibitor Screening
  • Cancer Research
  • Neurological Research
  • Drug Discovery

Features and Benefits

  • Efficient Procedure: Simplify your workflow with our hassle-free process, requiring only the addition of a single working reagent directly to your cell or lysate samples, saving valuable time and effort.
  • Compatibility with High-Throughput Systems: Seamlessly integrate our kit into high-throughput handling systems, ensuring smooth operations and accurate results, enhancing efficiency in your laboratory workflow.

Suitability

The kit is suitable for the quantitative determination of the apoptosis target caspase-3 activity and high throughput screening for apoptosis inducers and inhibitors in cell and tissue lysate.

Principle

The Caspase-3 Assay Kit provides a convenient means to measure caspase-3 activity in biological samples. In the assay, a specific substrate (N-Ac-DEVD-AFC) is cleaved by active caspase-3, forming a highly fluorescent product. The fluorescence intensity (λEx = 400 nm/λEm = 490 nm) is proportional to the caspase-3 activity.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is Caspase-3 Assay MAK457 compatible with flow cytometry for analysis? If yes, could you provide the mechanism or protocol for its use in flow cytometry? Thank you.

    1 answer
    1. The intended use for this kit is to assess Caspase-3 activity in a 96 well plate using a fluorometer. No specific protocol has been written for using this kit using a flow cytometer. Please contact your local TS team for specific product recommendations as it is likely there is an alternative caspase activity assay kit that has been optimized for this technique.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service