Skip to Content
Merck
All Photos(1)

Key Documents

NIST2391D

PCR-Based DNA Profiling Standard

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,650.00

$2,650.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$2,650.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,650.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 0.05 g

manufacturer/tradename

NIST®

technique(s)

PCR: suitable

application(s)

forensics and toxicology

storage temp.

2-8°C

General description

This PCR DNA profiling standard comprises well-characterized human deoxyribonucleic acid (DNA) in two forms, genomic DNA (Components A through D) and DNA to be extracted from cells that have been spotted onto Flinders technology associates (FTA) paper (Component E). The components are derived from human buffy coat white blood cells and labeled A, B, and C. Component E is a cell line obtained from American Type Culture Collection.

  • Component A contains 55 µL of single-source female genomic DNA in TE-4 buffer
  • Components B and C contain 55 µL each of single-source male genomic DNA in TE-4 buffer
  • Component D contains 55 µL of genomic DNA in TE-4 buffer which is a mix of components A and C in the ratio of 3:1
  • Component E contains single-source female cells spotted on two 6 mm punches of FTA paper such that each punch can hold approximately 7.5 × 104 cells
A unit of this standard reference material (SRM) is composed of one vial of each of the five components packaged together in a single box.

SRM 2391d_Cert


SRM 2391d_SDS

Application

This PCR DNA profiling standard is intended for use in the standardization of forensic and paternity quality assurance procedures for polymerase chain reaction (PCR) based genetic testing, for instructional law enforcement or non-clinical research purposes, and quality assurance when assigning values to in-house control materials.

Features and Benefits

  • The use of this material will provide confidence to the assignment of values to in-house prepared control materials for PCR DNA Profiling

Other Notes

  • Details on expiration, storage, safety, usage, and source are provided in the NIST certificate
  • Information on biomaterials, disposal, and transport is available in the SDS
  • The certified values for NIST2391d are derived from a combination of Capillary Electrophoresis (CE) and Next Generation Sequencing (NGS)-based characterizations that allowed for the counting and direct sequencing of short tandem repeats (STRs) at a locus.
Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Preparation Note

  • Before opening vials of components A, B, C, and D must be vortexed and centrifuged
  • Prior PCR amplification component E, cells on the FTA should be washed to remove PCR inhibitors and salts

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service