Skip to Content
Merck
All Photos(1)

Key Documents

NIST2372A

Human DNA quantitation standard

NIST® SRM® 2372a

Synonym(s):

NIST human DNA quantitation standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$2,610.00

$2,610.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 PKG
$2,610.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,610.00


Available to ship on15 April 2025Details


Request a Bulk Order

form

liquid

packaging

pkg of set (3 x 1 ea)

manufacturer/tradename

NIST®

technique(s)

PCR: suitable
qPCR: suitable

application(s)

genomic analysis

General description

This NIST DNA standard comprises three well-characterized human genomic DNA materials in pH 8.0 aqueous buffer. The components are derived from human buffy coat samples and labeled A, B, and C. Their composition is as follows:
  • Component A consists of genomic DNA from a single male donor.
  • Component B consists of genomic DNA from a single female donor.
  • Component C consists of a gravimetric mixture of genomic DNA (1 part male donor : 3 parts female donor).
A unit of the SRM consists of one sterile 0.5 mL vial of each component, each vial containing approximately 55 μL of DNA solution. Each of the vials is labeled and sealed with a color-coded screw cap.

SRM 2372a_cert

SRM 2372a _SDS

Application

Human DNA quantitation standard Standard Reference Material (SRM) is intended for use in the value assignment of human genomic deoxyribonucleic acid (DNA) forensic quantitation materials, primarily those used for quantitative polymerase chain reaction (qPCR).

Features and Benefits

  • SRM 2372a available with certificate documenting NIST-certified values of copy number & DNA mass concentration and non-certified values of mitochondrial genomes (mtDNA) relative to nuclear haploid genomes (nDNA)
  • Copy number values metrologically traceable to natural units and SI-derived units of volume
  • DNA mass concentration values metrologically traceable to natural units and SI-derived units of mass and volume
  • Assay-Specific mtDNA/nDNA Ratio values metrologically traceable to droplet digital polymerase chain reaction (ddPCR) measurement
  • Expiration details, and storage and use instructions provided on the NIST certificate
  • Notes on biomaterial handling, disposal, and transport documented in MSDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service