Skip to Content
Merck
All Photos(2)

Key Documents

NIST193

Potassium nitrate

NIST® SRM® 193

Sign Into View Organizational & Contract Pricing

Select a Size

90 G
$1,060.00

$1,060.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
90 G
$1,060.00

About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,060.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 90 g

manufacturer/tradename

NIST®

mp

334 °C (lit.)

application(s)

agriculture

format

neat

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM193 contains 90 g of highly purified and homogeneous crystalline potassium nitrate nitrogen.

SRM 193 _SDS

SRM 193 _CoA

Application

Potassium nitrate standard reference material (SRM) is intended for use as a working standard in determining the levels of potassium and nitrate nitrogen in fertilizer industry.

Features and Benefits

  • SRM 193 is available with a certificate documenting NIST-certified mass fraction values of potassium and nitrogen.
  • Certified values are determined using gravimetric assay.

Other Notes

Certified for the analytes listed below.
Potassium (K), Nitrogen (N)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service