Skip to Content
Merck
All Photos(1)

Key Documents

1.05065

Supelco

Potassium nitrate

99.995 Suprapur®, for inorganic trace analysis

Synonym(s):

Potassium nitrate, Nitric acid potassium salt

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$206.00
500 G
$1,380.00

$206.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
$206.00
500 G
$1,380.00

About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-818-8
NACRES:
NB.21
grade:
for inorganic trace analysis
form:
solid
solubility:
320 g/L

$206.00


Available to ship on07 April 2025Details


Request a Bulk Order

grade

for inorganic trace analysis

Quality Level

Assay

99.995%

form

solid

potency

3015 mg/kg LD50, oral (Rat)

pH

5.0-7.5 (20 °C, 50 g/L in H2O)

mp

334 °C

solubility

320 g/L

bulk density

800 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[N+](=O)([O-])[O-]

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Related Categories

Application


  • Effect of potassium nitrate on antioxidants production of date palm (Phoenix dactylifera L.) in vitro.: This study explores the role of potassium nitrate in enhancing antioxidant production in date palm cultures, indicating potential applications in plant tissue culture and agricultural biotechnology (Aldaej et al., 2014).

  • Bioflocculant production by Bacillus sp. Gilbert isolated from a marine environment in South Africa.: While the primary focus is on bioflocculant production, this research discusses the analysis and application potentials of potassium nitrate as a component in marine biotechnology, offering insights into microbial and environmental bioprocessing (Ugbenyen et al., 2014).

  • The scientific rationale and development of an optimized dentifrice for the treatment of dentin hypersensitivity.: This clinical study details the development of a dentifrice containing potassium nitrate, aimed at treating dentin hypersensitivity, showcasing the compound′s application in dental health products (Tavss et al., 2004).


Analysis Note

Purity (metallic): ≥ 99.995 %
Assay (acidimetric): ≥ 99.0 %
Chloride (Cl): ≤ 5 ppm
Sulfate (SO₄): ≤ 30 ppm
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 1.0 ppm
Ca (Calcium): ≤ 0.1 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.005 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.005 ppm
Cs (Cesium): ≤ 5 ppm
Cu (Copper): ≤ 0.005 ppm
Eu (Europium): ≤ 0.005 ppm
Fe (Iron): ≤ 0.05 ppm
Hg (Mercury): ≤ 0.01 ppm
La (Lanthanum): ≤ 0.005 ppm
Li (Lithium): ≤ 0.2 ppm
Mn (Manganese): ≤ 0.005 ppm
Mg (Magnesium): ≤ 0.10 ppm
Na (Sodium): ≤ 20 ppm
Ni (Nickel): ≤ 0.005 ppm
Pb (Lead): ≤ 0.005 ppm
Rb (Rubidium): ≤ 30 ppm
Sc (Scandium): ≤ 0.005 ppm
Sm (Samarium): ≤ 0.005 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.005 ppm
Y (Yttrium): ≤ 0.005 ppm
Yb (Ytterbium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.010 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service