Skip to Content
Merck
All Photos(1)

Key Documents

NIST1657

Synthetic refuse-derived fuel combustion calorimetric standard

NIST® SRM® 1657

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$709.00

$709.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$709.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$709.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

solid

packaging

pkg of 100 g

manufacturer/tradename

NIST®

application(s)

environmental

General description

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service