Skip to Content
Merck
All Photos(2)

Key Documents

169978

Sigma-Aldrich

2-Methoxybenzoic acid

ReagentPlus®, 99%

Synonym(s):

O-Methylsalicylic acid, o-Anisic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$42.40
100 G
$74.40

$42.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$42.40
100 G
$74.40

About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
509929
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$42.40


Check Cart for Availability

Request a Bulk Order

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

mp

98-100 °C (lit.)

functional group

carboxylic acid

SMILES string

COc1ccccc1C(O)=O

InChI

1S/C8H8O3/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

ILUJQPXNXACGAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxybenzoic acid was added as carbon supplement in the culture medium of Moraxella osloensis[1]. Photophysics of 2-methoxybenzoic acid has been investigated using both the time-correlated single photon counting and the fluorescence up-conversion techniques[2].

Application

2-Methoxybenzoic acid was used as internal standard during quantification of free and conjugated salicylic acid in tomato (Lycopersicon esculentum) cells by HPLC[3]. It was also employed in the synthesis of pthalides[4].

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

291.2 °F - closed cup

Flash Point(C)

144 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterocycles, 39, 47-47 (1994)
Involvement of endogenous salicylic acid content, lipoxygenase and antioxidant enzyme activities in the response of tomato cell suspension cultures to NaCl.
Molina A, et al.
The New phytologist, 156(3), 409-415 (2002)
The photophysics of salicylic acid derivatives in aqueous solution.
Pozdnyakov IP, et al.
Journal of the Physical Society of Japan, 22(5), 449-454 (2009)
R L Crawford et al.
Journal of bacteriology, 121(3), 794-799 (1975-03-01)
Gentisate:oxygen 1,2-oxidoreductase (decyclizing) (EC 1.13.11.4; gentisate 1,2-dioxygenase) from Moraxella osloensis was purified to homogeneity as shown by polyacrylamide gel electrophoresis. The enzyme has a molecular weight of about 154,000 and gives rise to subunits of molecular weight 40,000 in the
T Sasaki et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 50(5), 905-909 (1999-04-24)
For in vivo measurement of the hydroxyl radical (.OH), we synthesized [11C]salicylic acid, [11C]O-acetylsalicylic acid and [11C]2-methoxybenzoic acid by carboxylation of 2-bromomagnesiumanisol using [11C]CO2. The radiochemical yield of [11C]salicylic acid, [11C]O-acetylsalicylic acid and [11C]2-methoxybenzoic acid calculated from trapped [11C]CO2 in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service