Skip to Content
Merck
All Photos(2)

Key Documents

NIST1478

Polystyrene (narrow molecular weight distribution)

NIST® SRM® 1478

Synonym(s):

Polystyrene, PS

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$983.00

$983.00


Estimated to ship on25 March 2025


Request a Bulk Order

Select a Size

Change View
2 G
$983.00

About This Item

Linear Formula:
[CH2CH(C6H5)]n
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$983.00


Estimated to ship on25 March 2025


Request a Bulk Order

grade

certified reference material

Quality Level

form

beads

packaging

pkg of 2 g

manufacturer/tradename

NIST®

application(s)

chemicals and industrial polymers

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Polystyrene

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service