Skip to Content
Merck
All Photos(1)

Key Documents

M1875000

Mianserin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,2,3,4,10,14b-Hexahydro-2-methyl-dibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$218.00

$218.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$218.00

About This Item

Empirical Formula (Hill Notation):
C18H20N2 · HCl
CAS Number:
Molecular Weight:
300.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$218.00


Estimated to ship on16 April 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

mianserin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN1CCN2C(C1)c3ccccc3Cc4ccccc24

InChI

1S/C18H20N2.ClH/c1-19-10-11-20-17-9-5-3-7-15(17)12-14-6-2-4-8-16(14)18(20)13-19;/h2-9,18H,10-13H2,1H3;1H

InChI key

YNPFMWCWRVTGKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mianserin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Antidepressant; antagonist/inverse agonist at 5-HT2 serotonin receptors; also blocks the H1 histamine receptor and the α2 adrenoceptor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieyun Yin et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(9), G796-G801 (2014-03-15)
Mirtazapine is a noradrenergic and specific serotonergic antidepressant. The aim of this study was to investigate the effects of mirtazapine on gastrointestinal motility in dogs, including solid gastric emptying, antral and small intestinal contractions, and small intestinal and colonic transit.
Naveen Rajpurohit et al.
South Dakota medicine : the journal of the South Dakota State Medical Association, 67(4), 137-139 (2014-05-06)
We describe a rare case of severe drug-drug interaction between propafenone and mirtazapine leading to propafenone toxicity. A 69-year-old Caucasian male taking propafenone for atrial fibrillation was prescribed mirtazapine for insomnia. Subsequent to the first dose of mirtazapine the patient
Neeradi Dinesh et al.
Experimental parasitology, 144, 84-90 (2014-06-21)
In the present study, we have investigated the antileishmanial potential of mianserin, an antidepressant. Mianserin was found to inhibit both the promastigote and amastigote forms of the parasite in a dose dependant manner. The IC50 values for promastigotes and amastigotes
Maho Okubo et al.
Biochemical pharmacology, 93(1), 104-109 (2014-12-06)
A personalized treatment approach should be considered with the second-generation psychiatric drug mirtazapine because of high frequencies of side effects, including characteristic drowsiness. Plasma concentrations of mirtazapine in patients are influenced by many factors, including polymorphic cytochrome P450 enzymes contributing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service