Skip to Content
Merck
Mianserin hydrochloride British Pharmacopoeia (BP) Reference Standard

Mianserin hydrochloride

Mianserin hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1,2,3,4,10,14b-Hexahydro-2-methyl-dibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride
Empirical Formula (Hill Notation):
C18H20N2 · HCl
CAS Number:
Molecular Weight:
300.83
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing