IRMM523A
Aluminum
IRMM®, certified reference material, 0.1 mm foil
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL number:
UNSPSC Code:
11101705
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
IRMM®
form
foil
autoignition temp.
1400 °F
manufacturer/tradename
JRC
resistivity
2.6548 μΩ-cm
bp
2460 °C (lit.)
mp
660.37 °C (lit.)
density
2.7 g/mL at 25 °C (lit.)
format
matrix material
SMILES string
[Al]
InChI
1S/Al
InChI key
XAGFODPZIPBFFR-UHFFFAOYSA-N
Analysis Note
For more information please see:
IRMM523A
IRMM523A
Legal Information
IRMM is a registered trademark of European Commission
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[Conductive composite nanomaterial with biocompatible matrix and multilayer carbon nanotubes].
L P Ichkitidze et al.
Meditsinskaia tekhnika, (2)(2), 11-15 (2013-06-26)
Al18F: a new standard for radiofluorination.
David M Goldenberg et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(7), 1170-1170 (2013-05-21)
Seong-Hee Lee et al.
Journal of nanoscience and nanotechnology, 13(1), 509-512 (2013-05-08)
An ultrafine grain (UFG) complex lamella aluminum alloy sheet was successfully fabricated by ARB process using AA1050 and AA6061. The lamella thickness of the alloy became thinner and elongated to the rolling direction with increasing the number of ARB cycles.
Kate M Brougham et al.
Environmental pollution (Barking, Essex : 1987), 178, 89-96 (2013-04-03)
Although a great deal is known about the deposition of fluoride on vegetation, and the hazards associated with uptake by grazing herbivores, little is known about what happens to the concentration of fluoride in vegetation and soil at polluted sites
Andreza V Neri et al.
Anais da Academia Brasileira de Ciencias, 85(1), 87-102 (2013-03-30)
This study was carried out in Paraopeba National Reserve. It aims to classify and evaluate the soil of the studied area and to verify the influence of soil attributes on vegetation by testing the following hypotheses: 1) under woodland physiognomies
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service