Skip to Content
Merck
All Photos(1)

Key Documents

47288

Supelco

D-(+)-Maltose monohydrate

analytical standard

Synonym(s):

4-O-α-D-Glucopyranosyl-D-glucose, Maltobiose

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$54.30

$54.30


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 MG
$54.30

About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
93798
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

$54.30


Available to ship on16 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

feature

standard type disaccharide

analyte chemical class(es)

oligosaccharides

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-30°C

SMILES string

O.OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

D-(+)-Maltose monohydrate may be used as an analytical reference standard for the quantification of the analyte in honey samples using capillary gas chromatography.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of the capillary gas chromatographic analysis of mono-and oligosaccharides in honeys
Barez GAJ, et al.
Chromatographia, 50(7-8), 461-469 (1999)
S K Aoki et al.
Journal of bacteriology, 191(6), 1777-1786 (2009-01-07)
Contact-dependent growth inhibition (CDI) is a mechanism identified in Escherichia coli by which bacteria expressing two-partner secretion proteins encoded by cdiA and cdiB bind to BamA in the outer membranes of target cells and inhibit their growth. A third gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service