Skip to Content
Merck
All Photos(2)

Key Documents

F0750

Sigma-Aldrich

Fast Dark Blue R Salt

Solid

Synonym(s):

Azoic Diazo Component 51, Azoic Diazo No. 51

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$52.90

$52.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$52.90

About This Item

Empirical Formula (Hill Notation):
C14H10Cl2N5O4 · 0.5Cl4Zn
CAS Number:
Molecular Weight:
486.77
Colour Index Number:
37195
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$52.90


Check Cart for Availability

Request a Bulk Order

Product Name

Fast Dark Blue R Salt,

form

solid

Quality Level

composition

Dye content, ~30%

color

red-brown

solubility

ethanol: 10 mg/mL, dark red-brown

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Cl-].Cl[Zn]Cl.COc1cc([N+]#N)c(OC)cc1N=Nc2c(Cl)cc(cc2Cl)[N+]([O-])=O

InChI

1S/C14H10Cl2N5O4.3ClH.Zn/c1-24-12-6-11(13(25-2)5-10(12)18-17)19-20-14-8(15)3-7(21(22)23)4-9(14)16;;;;/h3-6H,1-2H3;3*1H;/q+1;;;;+2/p-3

InChI key

BHKNLVCPKUILSR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service