Skip to Content
Merck
All Photos(2)

Key Documents

364797

Sigma-Aldrich

Disperse Orange 3

Dye content 90 %

Synonym(s):

4-(4-Nitrophenylazo)aniline

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$131.00
25 G
$538.00

$131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$131.00
25 G
$538.00

About This Item

Linear Formula:
O2NC6H4N=NC6H4NH2
CAS Number:
Molecular Weight:
242.23
Colour Index Number:
11005
Beilstein:
1842907
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$131.00


Check Cart for Availability

Request a Bulk Order

composition

Dye content, 90%

mp

~200 °C (dec.) (lit.)

λmax

443 nm

SMILES string

Nc1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O

InChI

1S/C12H10N4O2/c13-9-1-3-10(4-2-9)14-15-11-5-7-12(8-6-11)16(17)18/h1-8H,13H2/b15-14+

InChI key

UNBOSJFEZZJZLR-CCEZHUSRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Orange 3 (DO3) is a monoazo dye, which contains 90% dye content plus dispersing agents and surfactants. It has two aromatic rings with an amino group on one of the rings and the other is substituted with a nitro group. It shows an absorption peak majoring at 415 nm.[1][2]

Application

DO3 can be used in low concentration to improve the electro-optic properties of dichroic polymer-dispersed liquid crystals, which can be used as materials for display devices.[3] It can also be used to functionalize epoxy-bisazo based polymers, which can be potentially used in sensors and optoelectronics.[4]

Features and Benefits

Used to provide experimental verification of a two-level model for the enhancement of the nonresonant, nonlinear refractive index, with a decrease in absorption by reduction of a donor group strength.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The role of molecular conformation and polarizable embedding for one-and two-photon absorption of disperse orange 3 in solution.
Silva DL, et al.
The Journal of Physical Chemistry B, 116(28), 8169-8181 (2012)
Biodegradation of a model azo disperse dye by the white rot fungus Pleurotus ostreatus.
Zhao X, et al.
International Biodeterioration & Biodegradation, 57(1), 1-6 (2006)
Analysis of dichroic dye-doped polymer-dispersed liquid crystal materials for display devices.
Kumar P, et al.
Thin Solid Films, 520(1), 457-463 (2011)
Epoxy-based polymers functionalized with bisazo chromophores: Synthesis, characterization and photoresponsive behavior.
Wang X, et al.
Polymer, 52(15), 3344-3356 (2011)
Anthony T-J Goon et al.
Contact dermatitis, 48(5), 248-250 (2003-07-19)
Cross-sensitization between para-phenylenediamine (PPD) and Disperse Orange 3 (DO3), among other textile dyes, has frequently been reported. We evaluated the frequency of simultaneous patch test reactions to PPD and a range of textile dyes. Retrospectively, we studied 128 patients who

Questions

  1. Is this dye a dichroic dye and mixable with liquid crystal ?

    1 answer
    1. Product 364797 is disperse orange 3. As per the literature as below, it is a dichroic dye and mixable with LCs.
      https://www.semanticscholar.org/paper/Induced-Alignment-in-Nematic-Liquid-Crystal-by-3-Sood-Raina/2f9f0f936ddf691bbfa727dd4d0b99fc0eabaa3f

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service