BCR457
Human thyroglobulin (Tg) (mass concentration)
BCR®, certified reference material
Synonym(s):
Thyroglobulin human
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
application(s)
clinical
pharmaceutical (small molecule)
format
matrix material
shipped in
wet ice
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
Analysis Note
For more information please see:
BCR457
BCR457
Legal Information
BCR is a registered trademark of European Commission
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Stine L Andersen et al.
European journal of endocrinology, 168(5), 723-731 (2013-02-28)
Placental transport of iodide is required for fetal thyroid hormone production. The sodium iodide symporter (NIS) mediates active iodide transport into the thyroid and the lactating mammary gland and is also present in placenta. NIS is competitively inhibited by thiocyanate
Andrew N Hoofnagle et al.
The Journal of clinical endocrinology and metabolism, 98(4), 1343-1352 (2013-03-02)
Serum thyroglobulin (Tg) measurements are central to the management of patients treated for differentiated thyroid carcinoma. For decades, Tg measurements have relied on methods that are subject to interference by commonly found substances in human serum and plasma, such as
Mark M Kushnir et al.
Clinical chemistry, 59(6), 982-990 (2013-02-12)
Measurement of serum thyroglobulin (Tg) is used to monitor patients after treatment for differentiated thyroid carcinoma (TC). Difficulty in using Tg as a biomarker of the recurrence of TC in many patients stems from the presence of endogenous anti-Tg autoantibodies
Prognostic significance of FDG PET/CT on the follow-up of patients of differentiated thyroid carcinoma with negative I131 whole- body scan and elevated thyroglobulin levels.
Carol S Marcus
Clinical nuclear medicine, 38(3), 195-195 (2013-01-29)
Fiona L R Williams et al.
The Journal of clinical endocrinology and metabolism, 98(2), 829-838 (2013-01-17)
Relatively little is known in euthyroid populations about the changes in maternal thyroid hormones during pregnancy, the nature of the relationship to cord thyroid hormone levels, and subsequent infant neurodevelopment. The aim of the study was to describe the relationship
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service