Skip to Content
Merck
All Photos(1)

Key Documents

609312

Sigma-Aldrich

Thyroglobulin, Human Thyroid

Synonym(s):

Thyroglobulin, Human Thyroid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$205.00

$205.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$205.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

$205.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

human thyroid

Quality Level

Assay

≥95% (PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

2-8°C

General description

Native thyroglobulin, consisting of two polypeptide monomers of 330 kDa each, purified from human thyroid. This protein is an excellent source of N-linked glycans of various structures, including sialylated core fucose and oligomannose oligosaccharides. Also, Gal-3-SO4 and GalNAc-6-SO4 residues located in a single multi-branched complex type glycan have been reported in calf thymus and human proteins. When subjected to native PAGE this protein shows a band at

~660 kDa, representing the dimeric form, and another band at ~330 kDa, representing the monomeric form.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 20 mM NH₄HCO₃.

Preparation Note

Prepared from tissues of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Reconstitute in phosphate buffer, containing 150 mM NaCl, pH >7.0.

Other Notes

Rawitch, A.B., et al. 1993. Arch. Biochem. Biophys. 300, 271.
Black, E.G., and Sheppard, M.C. 1991. Clin. Endocrinol. 35, 519.
Spiro, R.G., and Bhoyroo, V.D. 1988. J. Biol. Chem. 263, 14351.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service