Skip to Content
Merck
All Photos(1)

Key Documents

BCR114

Potassium sulfate fertilizer (elemental composition)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$381.00

$381.00


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
100 G
$381.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$381.00


Estimated to ship on28 March 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

agriculture
environmental

format

matrix material

Analysis Note

For more information please see:
BCR114

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service