Skip to Content
Merck
All Photos(1)

Documents

B2551

Millipore

Brewer thioglycollate medium

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Thioglycollate Medium (Brewer)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

pH

7.2±0.2 (25 °C)

application(s)

clinical testing
food and beverages
sterility testing

microbiology

suitability

anaerobic bacteria
nonselective for Bacteroides spp.
nonselective for Clostridium spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)

Related Categories

Application

Used for testing the sterility of biological products.

Components

Components (g/L)
Proteose Peptone 10.00
Infusion from Beef 17.50
Dextrose 5.00
Sodium Chloride 5.00
Dipotassium Phosphate 2.00
Sodium Thioglycollate 0.50
Methylene Blue 0.002
Agar 0.50

Preparation Note

Suspend 40.5 g of Brewer Thioglycollate Medium in 1000 ml of distilled water. Boil to dissolve the medium completely. Dispense in tubes or in suitable containers as desired and sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes.Note: If more than the upper one third of medium has acquired a green color, the medium may be restored once by heating on a water bath or free flowing steam until the green color disappears.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin R Kingery et al.
Basic research in cardiology, 112(2), 19-19 (2017-02-27)
In the failing heart, iNOS is expressed by both macrophages and cardiomyocytes. We hypothesized that inflammatory cell-localized iNOS exacerbates left ventricular (LV) remodeling. Wild-type (WT) C57BL/6 mice underwent total body irradiation and reconstitution with bone marrow from iNOS
Xuyun Gu et al.
Experimental and therapeutic medicine, 16(1), 171-177 (2018-06-14)
Sepsis with severe systemic inflammation remains a great challenge for the intensive care unit in clinics. Although biomarkers have been identified to diagnose, monitor and predict these syndromes, novel therapeutic approaches are required for the amelioration of symptoms of sepsis
F Castro et al.
British journal of cancer, 101(8), 1329-1337 (2009-10-15)
Elderly individuals react less efficiently to vaccines than do adults, mainly because of T-cell unresponsiveness. In this study, we analysed whether tumour-associated antigen (TAA)-specific CD8 T-cell responses could be induced by vaccination in old mice with metastatic breast cancer. The
National Institutes of Health Bulletin (1941)
Thorsten Wichmann et al.
Archives of orthopaedic and trauma surgery, 139(4), 451-459 (2018-11-09)
Knitted cotton outer gloves offer protection against surgical glove perforation and provide improved grip on instruments. These gloves absorb blood and other fluids during surgery, and may therefore also accumulate contaminating bacteria. To date, there is no published data on

Articles

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service