Skip to Content
Merck
All Photos(1)

Key Documents

A7472

Sigma-Aldrich

Allophycocyanin

suspension, 4 mg/mL

Synonym(s):

APC

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
$795.00

$795.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.5 ML
$795.00

About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

$795.00


Check Cart for Availability

Request a Bulk Order

form

suspension

Quality Level

mol wt

104 kDa

composition

Suspension in 60% saturated Ammonium Sulfate and 50 mM Potassium Phosphate, pH 7.0

concentration

4 mg/mL

technique(s)

microbe id | staining: suitable

color

blue

λmax

645-655 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

Phycobiliprotein exhibiting exceptional absorbance and fluorescence characteristics due to a lack of susceptibility to internal and external fluorescence quenching. Ideal for highly sensitive studies in flow cytometry and immunoassays.

Biochem/physiol Actions

Allophycocyanin is a pigment which allows the energy trapped by phycobiliproteins to be transferred to the chloroplast lamellae.[1]

Physical form

4 mg/mL solution in 60% saturated ammonium sulfate and 50 mM potassium phosphate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Energy transfer in phycobilisomes from phycoerythrin to allophycocyanin.
Gantt E and Lipschultz CA
Biochimica et Biophysica Acta, 292, 858-861 (1973)
Margit Kaldmäe et al.
Protein science : a publication of the Protein Society, 28(6), 1024-1030 (2019-03-31)
Biotechnological applications of protein complexes require detailed information about their structure and composition, which can be challenging to obtain for proteins from natural sources. Prominent examples are the ring-shaped phycoerythrin (PE) and phycocyanin (PC) complexes isolated from the light-harvesting antennae
Erica D'Aguanno et al.
Life (Basel, Switzerland), 5(1), 969-996 (2015-03-21)
How did primitive living cells originate? The formation of early cells, which were probably solute-filled vesicles capable of performing a rudimentary metabolism (and possibly self-reproduction), is still one of the big unsolved questions in origin of life. We have recently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service