Skip to Content
Merck
All Photos(1)

Key Documents

A1200000

Anhydrotetracycline hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(4S,4aS,12aS)-4-(Dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$265.00

$265.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
$265.00

About This Item

Empirical Formula (Hill Notation):
C22H22N2O7 · HCl
CAS Number:
Molecular Weight:
462.88
Beilstein:
4228856
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$265.00


Available to ship on15 April 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

tetracycline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].[H][C@@]12Cc3c(C)c4cccc(O)c4c(O)c3C(=O)[C@]1(O)C(=O)C(C(N)=O)=C(O)[C@H]2N(C)C

InChI

1S/C22H22N2O7.ClH/c1-8-9-5-4-6-12(25)13(9)17(26)14-10(8)7-11-16(24(2)3)18(27)15(21(23)30)20(29)22(11,31)19(14)28;/h4-6,11,16,25-27,31H,7H2,1-3H3,(H2,23,30);1H/t11-,16-,22-;/m0./s1

InChI key

XBSQEFHDCDFNJU-WPJNXPDPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Anhydrotetracycline hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan W Kotula et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(13), 4838-4843 (2014-03-19)
The mammalian gut is a dynamic community of symbiotic microbes that interact with the host to impact health, disease, and metabolism. We constructed engineered bacteria that survive in the mammalian gut and sense, remember, and report on their experiences. Based
Emad M Hussien
Biomedical chromatography : BMC, 28(9), 1278-1283 (2014-03-13)
A novel reversed-phase HPLC method was developed and validated for the assay of tetracycline hydrochloride and the limit of 4-epianhydrotetracycline hydrochloride impurity in tetracycline hydrochloride commercial bulk and pharmaceutical products. The method employed L1 (3 µm, 150 × 4.6 mm) columns, a mobile phase
Eric M Ransom et al.
Applied and environmental microbiology, 81(5), 1652-1660 (2014-12-21)
Fluorescent proteins are powerful reporters in biology, but most require O2 for chromophore maturation, making them inherently difficult to use in anaerobic bacteria. Clostridium difficile, a strict anaerobe with a genomic GC content of only 29%, is the leading cause
Sebastiaan Werten et al.
Biochemistry, 53(50), 7990-7998 (2014-11-29)
Genes that render bacteria resistant to tetracycline-derived antibiotics are tightly regulated by repressors of the TetR family. In their physiologically relevant, magnesium-complexed form, tetracyclines induce allosteric rearrangements in the TetR homodimer, leading to its release from the promoter and derepression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service