Merck
  • Development and validation of an HPLC method for tetracycline-related USP monographs.
Development and validation of an HPLC method for tetracycline-related USP monographs.
Biomedical chromatography : BMC (2014-03-13)
Emad M Hussien
ABSTRACT
A novel reversed-phase HPLC method was developed and validated for the assay of tetracycline hydrochloride and the limit of 4-epianhydrotetracycline hydrochloride impurity in tetracycline hydrochloride commercial bulk and pharmaceutical products. The method employed L1 (3 µm, 150 × 4.6 mm) columns, a mobile phase of 0.1% phosphoric acid and acetonitrile at a flow rate of 1.0 mL/min, and detection at 280 nm. The separation was performed in HPLC gradient mode. Forced degradation studies showed that tetracycline eluted as a spectrally pure peak and was well resolved from its degradation products. The fast degradation of tetracycline hydrochloride and 4-epianhydrotetracycline hydrochloride in solution was retarded by controlling the autosampler temperature at 4 °C and using 0.1% H3 PO4 as diluent. The robustness of the method was tested starting with the maximum variations allowed in the US Pharmacopeia (USP) general chapter Chromatography <621>. The method was linear over the range 80-120% of the assay concentration (0.1 mg/mL) for tetracycline hydrochloride and 50-150% of the acceptance criteria specified in the individual USP monographs for 4-epianhydrotetracycline hydrochloride. The limit of quantification for 4-epianhydrotetracycline hydrochloride was 0.1 µg/mL, 20 times lower than the acceptance criteria. The method was specific, precise, accurate and robust.
Product Description
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Anhydrotetracycline hydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Anhydrotetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Tetracycline hydrochloride, powder
Tetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Tetracycline hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Anhydrotetracycline hydrochloride, VETRANAL®, analytical standard
Supelco
4-Epianhydrotetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material