Skip to Content
Merck
All Photos(2)

Key Documents

96996

Supelco

Biphenyl

certified reference material, TraceCERT®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H5
CAS Number:
Molecular Weight:
154.21
Beilstein:
1634058
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E230
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

5.31 (vs air)

vapor pressure

9.46 mmHg ( 115 °C)

product line

TraceCERT®

autoignition temp.

1004 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

0.6 %, 111 °F
5.8 %, 166 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

255 °C (lit.)

mp

68-70 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

c1ccc(cc1)-c2ccccc2

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jiyoung Seo et al.
Applied microbiology and biotechnology, 91(2), 219-228 (2011-06-01)
This review details recent progresses in the flavonoid biotransformation by bacterial non-heme dioxygenases, biphenyl dioxygenase (BDO), and naphthalene dioxygenase (NDO), which can initially activate biphenyl and naphthalene with insertion of dioxygen in stereospecfic and regiospecific manners. Flavone, isoflavone, flavanone, and
Pasha M Khan et al.
Bioorganic & medicinal chemistry letters, 23(2), 532-536 (2012-12-13)
The structure-activity relationship study of a diphenylpropanamide series of ROR-γ selective modulators is reported. Compounds were screened using chimeric receptor Gal4 DNA-binding domain (DBD)-NR ligand binding domain cotransfection assay in a two-step format. Three different regions of the scaffold were
Kathy D Miller et al.
Breast cancer research and treatment, 148(1), 99-106 (2014-09-27)
Preclinical data suggest that medroxyprogesterone acetate (MPA) has both anti-metastatic and anti-angiogenic activity in the absence of hormone receptors (HR). This phase II trial assessed the activity of MPA alone or in combination with low-dose chemotherapy in patients with metastatic
Yang Ou et al.
Chemical communications (Cambridge, England), 49(33), 3473-3475 (2013-03-22)
A novel A21-CuI catalyzed direct nitrogenation of biphenyl halides for the direct synthesis of carbazoles via a direct C-H amination process has been developed. A recyclable and inexpensive Cu-catalyst was successfully employed in N-heterocyclic compound synthesis via tandem azidation and
Xiaofeng Chen et al.
The Journal of organic chemistry, 77(17), 7572-7578 (2012-08-07)
A highly efficient cross-coupling of diarylborinic acids and anhydrides with aryl chlorides and bromides has been effected by using a palladium catalyst system co-supported by a strong σ-donor N-heterocyclic carbene (NHC), N,N'-bis(2,6-diisopropylphenyl) imidazol-2-ylidene, and a strong π-acceptor phosphite, triphenylphosphite, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service