Skip to Content
Merck
All Photos(1)

Documents

93159

Supelco

Multi Cation Standard 2 for IC

TraceCERT®, ≤0.1% in nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

composition

Ba, 100 mg/L
Ca, 100 mg/L
K, 100 mg/L
Li, 100 mg/L
Mg, 100 mg/L
Mn, 100 mg/L
Na, 100 mg/L
NH4, 100 mg/L
Sr, 100 mg/L

concentration

≤0.1% in nitric acid

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

multi-component solution

Linkage

Visit the IC Portal to learn more

Analysis Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service