Skip to Content
Merck
All Photos(1)

Key Documents

62414

Supelco

Sodium bicarbonate/Sodium carbonate concentrate

Sodium bicarbonate/Sodium carbonate concentrate

Na2CO3 64mM and NaHCO3 20mM in water, IC eluent concentrate (20x) for Metrosep A Supp 5

Synonym(s):

Buffer Solution Concentrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$166.00
2.5 L
$302.00

$166.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$166.00
2.5 L
$302.00

About This Item

UNSPSC Code:
12161700
NACRES:
NB.21

$166.00


Available to ship on07 April 2025Details


Request a Bulk Order

Quality Level

concentration

20 mM NaHCO3 in H2O
64 mM Na2CO3 in H2O

technique(s)

ion chromatography: suitable

suitability

application tested

General description

The “bicarbonate concentrate” is 1-molar solution of sodium bicarbonate (the saturation temperature of the 1-molar solution is 15°C). It is highly advantageous as it is temperature dependent and its subsequent dilution with water can be appropriately controlled.[1]
This eluent concentrate is application tested. Information about the exact contents and expiry date is described in the certificate.

Application

Metrohm IC application note AN-S-331 :Separation of seven standard anions with the Metrosep A Supp 5 microbore anion column.

Metrohm IC application note AN-S-313 :Dual detection of nitrite traces in the presence of excess chloride concentrations with the aid of the 940 Professional IC Vario (conductivity, UV/VIS).

Metrohm IC application note AN-S-309 :Anions in 70% hydrogen peroxide applying Inline Matrix Elimination.

Metrohm IC application note AN-S-308 : Azide in air from pharmaceutical production.

Metrohm IC application note AN-S-304 :Variable Inline Preconcentration including Matrix Elimination for trace anion analysis (MiPCT-ME)

Metrohm IC application note AN-S-290 :Chlorine, bromine, and sulfur in low-density polyethylene (ERM-EC680k) applying Combustion Ion Chromatographhy (CIC).

Linkage

Visit the IC Portal to learn more

Preparation Note

Prepared with sodium carbonate, sodium bicarbonate and high purity water (18.2 MΩ, 0.2 μm filtered)

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cardiovascular Disorders in Hemodialysis.
Claudio Ronco, Alessandra Brendolan, Nathan W. Levin.
Science, 20-22 (2005)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service