Skip to Content
Merck
All Photos(1)

Key Documents

93009

Millipore

E.coli Chromogenic ID Membrane

suitable for microbiology, white colored, sterile, diameter 70 mm

Synonym(s):

Indicator membrane, E. coli ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
$132.00

$132.00


Estimated to ship on22 May 2025


Request more information

Select a Size

Change View
25 DISCS
$132.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

$132.00


Estimated to ship on22 May 2025


Request more information

membrane size

70 mm

sterility

sterile

form

disc

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

agriculture
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

General description

E. coli chromogenic ID Membranes are substrate-carrying membranes used for rapid identification and differentiation of E. coli through characteristic chromogenic reactions while placed on the agar surface post-incubation. The membranes contain chromogenic substrate X-Gal. The principle is based on the ability of E. coli to cleave X-Gal substrate, giving a blue colony that is visible post-incubation.

Application

E. coli Chromogenic ID Membrane is used for rapid detection & confirmation of Escherichia coli in water and food samples, based on a chromogenic method. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Pseudomonas aeruginosa (27853) - Colorless

Escherichia coli (25922) - BlueStaphylococcus aureus (25923) - ColorlessSalmonella typhimurium (14028) - Colorless

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service