90232
Color Reference Solutions
related to Ph.Eur Color intensity testing, analytical standard, Set of colors B, BY, Y, GY, R
Synonym(s):
Color Calibration Standards
About This Item
Recommended Products
grade
analytical standard
Quality Level
Agency
according to Ph. Eur.
shelf life
limited shelf life, expiry date on the label
packaging
ampule of 10 mL
application(s)
cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical
format
multi-component solution
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Ready-to-use solutions
- Prepared and qualified in accordance with Ph. Eur.
- Available in tamper-evident 10 mL ampules
- Expiry dated on the label with a limited shelf life
- Accompanied with a detailed certificate of analysis for an effective quality management system
Components
- Color Reference Solutions B acc. to PhEur - 10 X 10mL ampules
- Color Reference Solutions BY acc. to PhEur - 8 X 10mL ampules
- Color Reference Solutions Y acc. to PhEur - 8 X 10mL ampules
- Color Reference Solutions GY acc. to PhEur - 8 X 10mL ampules
- Color Reference Solutions R acc. to PhEur - 8 X 10mL ampules
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Carc. 1B - Met. Corr. 1 - Repr. 1B
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service