Skip to Content
Merck
All Photos(1)

Key Documents

86293

Supelco

Color Reference Solutions

related to Ph.Eur Color intensity testing, analytical standard, Set of colors BY

Synonym(s):

Color calibration standard brownish yellow (BY1 to BY7)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$495.00

$495.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$495.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$495.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Agency

according to Ph. Eur.

shelf life

limited shelf life, expiry date on the label

packaging

ampule of 2 mL

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

The kit contains a set of seven brownish yellow color reference standards (identified sequentially from BY1 to BY7) and a 3.7% hydrogen chloride solution in water. The solutions are produced from three primary metal salt solutions of ferric(III) chloride, cobalt(II) chloride, and copper(II) sulfate pentahydrate, in accordance with chapter 2.2.2 ′Degree of colouration of liquids′ of the European Pharmacopoeia (Ph. Eur.).

Application

The reference standards are used as calibration and/or quality control standards to measure the degree of coloration of liquids in the brown-yellow-red color range.

Features and Benefits

  • Ready-to-use solutions
  • Prepared and qualified in accordance with Ph. Eur.
  • Available in tamper-evident 2 mL ampules
  • Expiry dated on the label with a limited shelf life
  • Accompanied with a detailed certificate of analysis for an effective quality management system

Components

This is a set of the following color reference solutions according to PhEur:

  • Colour Reference Solution BY1 - 2mL ampule
  • Colour Reference Solution BY2 - 2mL ampule
  • Colour Reference Solution BY3 - 2mL ampule
  • Colour Reference Solution BY4 - 2mL ampule
  • Colour Reference Solution BY5 - 2mL ampule
  • Colour Reference Solution BY6 - 2mL ampule
  • Colour Reference Solution BY7 - 2mL ampule
  • Blind sample : Hydrochloric acid solution, 3.7% in water - 2mL ampule

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Met. Corr. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service