Skip to Content
Merck
All Photos(2)

Key Documents

901812

Sigma-Aldrich

FixCat PF6

≥95%

Synonym(s):

[1,3-Bis(2,6-diisopropylphenyl)-4-[(4-ethyl-4-methylpiperazinium-1-yl)methyl]-2-imidazolidinylidene]dichloro(2-isopropoxybenzylidene)ruthenium(II) hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H67Cl2F6N4OPRu
CAS Number:
Molecular Weight:
996.98
UNSPSC Code:
12352101

Assay

≥95%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

storage temp.

2-8°C

Application

FixCat PF6 is highly active for heterogeneous olefin metathesis. The catalyst can be used in metathesis of water-insoluble substrates in ′′classical′′ organic solvents and can be used in reactions in ionic liquids. FixCat PF6 is efficient in RCM (ring closing metathesis) macrocyclization in more concentrated reaction mixtures.

Learn more about our metathesis catalysts

Legal Information

Sold in collaboration with Apeiron Synthesis, Inc. Research quantities are 100 g / customer / year. For quantities that exceed the research limit, please contact the supplier at www.apeiron-synthesis.com. PCT/EP2013/053967

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service