Skip to Content
Merck
All Photos(4)

Key Documents

774901

Sigma-Aldrich

Grubbs Catalyst® M101

Umicore

Synonym(s):

Grubbs Catalyst® M1, Dichloro(3-phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$162.00
5 G
$751.00

$162.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$162.00
5 G
$751.00

About This Item

Empirical Formula (Hill Notation):
C51H76Cl2P2Ru
CAS Number:
Molecular Weight:
923.07
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$162.00


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

120-141 °C

storage temp.

2-8°C

SMILES string

Cl[Ru](Cl)(=C1C=C(c2ccccc2)c3ccccc13)([PH](C4CCCCC4)(C5CCCCC5)C6CCCCC6)[PH](C7CCCCC7)(C8CCCCC8)C9CCCCC9

InChI

1S/2C18H33P.C15H10.2ClH.Ru/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-6-12(7-3-1)15-11-10-13-8-4-5-9-14(13)15;;;/h2*16-18H,1-15H2;1-9,11H;2*1H;

InChI key

QIRGCWAEHCIFAL-UHFFFAOYSA-N

Application

Metathesis Catalyst

Legal Information

Product of Umicore

Additional information available at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service