Skip to Content
Merck
All Photos(1)

Key Documents

89921

Supelco

(±)-2,4-Dihydroxybutyric acid lithium salt

≥95.0% (GC)

Synonym(s):

2,4-Dihydroxybutanoic acid lithium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$323.00
50 MG
$646.00

$323.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$323.00
50 MG
$646.00

About This Item

Empirical Formula (Hill Notation):
C4H8O4
CAS Number:
Molecular Weight:
120.10
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

$323.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0% (GC)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(CCO)C(=O)O

InChI

1S/C4H8O4/c5-2-1-3(6)4(7)8/h3,5-6H,1-2H2,(H,7,8)

InChI key

UFYGCFHQAXXBCF-UHFFFAOYSA-N

Biochem/physiol Actions

Various cases of succinic semialdehyde dehydrogenase deficiency have shown consistently increased amounts of this metabolite, while 2,4-dihydroxybutanoic acid is usually absent in normal human urine extracts or in only trace constituents in neonates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minna Hartonen et al.
Journal of chromatography. A, 1293, 142-149 (2013-05-07)
Comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry (GC×GC-TOFMS) was applied in the quantification and identification of organic compounds in patient-matched human cerebrospinal fluid (CSF) and serum samples. Concentrations of 21 amino and hydroxyl acids varied from 0.04 to 77ng/μl in CSF
M Orešič et al.
Translational psychiatry, 1, e57-e57 (2011-01-01)
Mild cognitive impairment (MCI) is considered as a transition phase between normal aging and Alzheimer's disease (AD). MCI confers an increased risk of developing AD, although the state is heterogeneous with several possible outcomes, including even improvement back to normal
G K Brown et al.
Journal of inherited metabolic disease, 10(4), 367-375 (1987-01-01)
In addition to the previously reported abnormalities, urine extracts from three cases of succinic semialdehyde dehydrogenase deficiency have shown consistently increased amounts of 2,4-dihydroxybutyric acid, and its lactone, and 3-hydroxypropionic acid, metabolites related to the alpha-oxidation of 4-hydroxybutyric acid. Threo-
J A Thompson et al.
Clinical chemistry, 21(13), 1892-1898 (1975-12-01)
Gas chromatographic/mass spectrometric/computer analyses of trimethylsilyl derivatives of urine extracts, prepared by an anion-exchange procedure, reveal the presence of several tetronic and deoxytetronic acids. Identifications were confirmed by comparing gas-chromatographic methylene unit values and mass spectrometric characteristics with those of
Bobbie-Jo Webb-Robertson et al.
Metabolomics : Official journal of the Metabolomic Society, 10(5), 897-908 (2014-09-26)
Urease pre-treatment of urine has been utilized since the early 1960s to remove high levels of urea from samples prior to further processing and analysis by gas chromatography-mass spectrometry (GC-MS). Aside from the obvious depletion or elimination of urea, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service