Skip to Content
Merck
All Photos(1)

Key Documents

76046

Supelco

Piperidine

analytical standard

Synonym(s):

Hexahydropyridine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$86.00

$86.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$86.00

About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
Beilstein:
102438
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$86.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

3 (vs air)

vapor pressure

23 mmHg ( 20 °C)

Assay

≥98.0% (GC)
98.0-102.0% (T)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.452 (lit.)

bp

106 °C (lit.)

mp

−13 °C (lit.)

density

0.862 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C1CCNCC1

InChI

1S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2

InChI key

NQRYJNQNLNOLGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Phillip Turner et al.
ACS chemical neuroscience, 5(7), 552-558 (2014-04-03)
Alzheimer's disease (AD) is the most common form of dementia and the sixth leading cause of death in the United States. Plaques composed of aggregated amyloid-beta protein (Aβ) accumulate between the neural cells in the brain and are associated with
P Ricardo Girling et al.
Organic & biomolecular chemistry, 9(9), 3105-3121 (2011-03-11)
A review into the aza-Diels-Alder reaction, mainly concentrating on literature examples that form piperidin-4-ones from the reaction of imines and electron rich dienes or enones, either through a Lewis acidic/Brønsted acid approach or through the use of an organocatalyst. This
Ryo Hayashi et al.
Bioorganic & medicinal chemistry, 22(15), 3824-3828 (2014-07-19)
While stimulation of formyl peptide receptors (FPRs) on the surface of human neutrophils induces several immune responses, under conditions of continuous activation of the receptor by agonists such as formyl-Met-Leu-Phe-OH (fMLP), neutrophil-dependent tissue damage ensues. Thus, FPR antagonists could be
Rachel Stephenson et al.
Nanomedicine : nanotechnology, biology, and medicine, 10(8), 1799-1808 (2014-07-12)
Gonadotropin-releasing hormone (GnRH) receptors are overexpressed on many cancer cells but not on primary cell lines. This study was designed to investigate the targeting ability and uptake of dendritic lipidated [Gln(1)]-GnRH peptide analogues on receptor-positive prostate cancer PC-3 cells relative
Lars Erik Uggerhøj et al.
FEBS letters, 588(17), 3291-3297 (2014-07-27)
The inclusion of peptoid monomers into antimicrobial peptides (AMPs) increases their proteolytic resistance, but introduces conformational flexibility (reduced hydrogen bonding ability and cis/trans isomerism). We here use NMR spectroscopy to answer how the insertion of a peptoid monomer influences the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service