Skip to Content
Merck
All Photos(1)

Key Documents

86903

Supelco

Tetrabutylammonium iodide

suitable for ion pair chromatography, LiChropur, ≥99.0%

Synonym(s):

TBAI

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$341.00

$341.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$341.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(I)
CAS Number:
Molecular Weight:
369.37
Beilstein:
3916152
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

$341.00


Check Cart for Availability

Request a Bulk Order

description

cationic

Quality Level

Assay

≥99.0% (AT)
≥99.0%

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

mp

141-143 °C (lit.)
144-148 °C

λ

10 % in acetonitrile

UV absorption

λ: 290 nm Amax: 0.1
λ: 300 nm Amax: 0.05
λ: 320 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[I-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.HI/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

DPKBAXPHAYBPRL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium iodide is an ion-pair chromatography (IPC) reagent suitable for anionic separation sorted by carbon chain length.[1]

Application

Tetrabutylammonium iodide may be used as a mobile phase additive in ion-pair high-performance liquid chromatography (IP-HPLC) assay of 4-aminopyridine in serum.[1] Tetrabutylammonium iodide may also be used as a mobile phase additive in the analysis of tetracycline by reversed-phase IPC. The addition of tetrabutylammonium iodide regulates the retention of tetracyclines.[2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ion-pair high-performance liquid chromatographic assay of 4-aminopyridine in serum
Shinohara Y, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 230(2), 363-372 (1982)
Reversed-phase ion-pair liquid chromatography of tetracyclines
Eksborg S and Ekqvist B
Journal of Chromatography A, 209(2), 161-173 (1981)
Chao Zhong et al.
Carbohydrate polymers, 94(1), 38-45 (2013-04-03)
In this article, a novel and high efficient solvent, tetra-n-Butylammonium Hydroxide (TBAH), was used for dissolution and isolation of straw cellulose from wheat straw. The composition analysis with gas chromatography (GC) and the spectroscopic characterization analysis conducted by X-Ray diffraction
Fabio Galeotti et al.
Journal of chromatography. A, 1284, 141-147 (2013-03-05)
In this study, by using tetrabutylammonium bisulfate as ion-pairing reagent, we were able to separate all the main heparin/heparan sulfate disaccharides generated by the action of heparinases along with the main Hep tetrasaccharide possessing a 3-O-sulfate group on the sulfoglucosamine
Erbo Shi et al.
Organic letters, 14(13), 3384-3387 (2012-06-27)
A metal-free C-H oxidation for the construction of allylic esters has been developed. The use of a commercially available and inexpensive catalyst and oxidant, and readily available starting materials, coupled with the operational simplicity of the reaction, renders the methodology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service