Skip to Content
Merck
All Photos(2)

Key Documents

861235

Sigma-Aldrich

Methyl Red hydrochloride

ACS reagent

Synonym(s):

4-Dimethylaminoazobenzene-2′-carboxylic acid hydrochloride, Acid Red 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$65.40

$65.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$65.40

About This Item

Linear Formula:
(CH3)2NC6H4N=NC6H4CO2H·HCl
CAS Number:
Molecular Weight:
305.76
Colour Index Number:
13020
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$65.40


Check Cart for Availability

Request a Bulk Order

grade

ACS reagent

Quality Level

form

powder or crystals

visual transition interval

4.2-6.2, pink to yellow

mp

175 °C (dec.) (lit.)

solubility

ethanol: 0.1%

λmax

493 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.CN(C)c1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C15H15N3O2.ClH/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;/h3-10H,1-2H3,(H,19,20);1H/b17-16+;

InChI key

ICTWKXAVJAHRMK-CMBBICFISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl red (C.I. 13020) is also known as acid red 2. The hydrochloride form is slightly soluble in water and ethanol.

Application

Methyl red is widely used in food science and applied & pure biology. It is added in media while preparing agar plates for microbiology. It is used as a vital stain for protozoa. Methyl red is also utilized as a tracer for assessing penetration of foods into packaging and as a component of an indicator to examine pork quality. It has been used to study the ability of earthworms to modify their environmental pH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service