Skip to Content
Merck
All Photos(4)

Key Documents

82704

Sigma-Aldrich

Pyridine

puriss., absolute, over molecular sieve (H2O ≤0.005%), ≥99.8% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

grade

absolute
puriss.

Assay

≥99.8% (GC)

autoignition temp.

899 °F

quality

over molecular sieve (H2O ≤0.005%)

expl. lim.

12.4 %

impurities

≤0.005% water
water

refractive index

n20/D 1.509 (lit.)

bp

115 °C (lit.)

mp

−42 °C (lit.)

solubility

water: soluble

density

0.978 g/mL at 25 °C (lit.)

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porous monodisperse V2O5 microspheres as cathode materials for lithium-ion batteries.
Wang S, et al.
Journal of Materials Chemistry, 21(17), 6365-6369 (2011)
Metal-organic frameworks based on iron oxide octahedral chains connected by benzenedicarboxylate dianions.
Whitfield TR, et al.
Solid State Sciences, 7(9), 1096-1103 (2005)
The far-infrared spectra of metal-halide complexes of pyridine and related ligands.
Clark RJH and Williams CS.
Inorganic Chemistry, 4(3), 350-357 (1965)
An investigation of Knoevenagel condensation reaction in microreactors using a new zeolite catalyst.
Zhang X, et al.
Applied Catalysis A: General, 261(1), 109-118 (2004)
Total syntheses of complanadines A and B.
Le Zhao et al.
Angewandte Chemie (International ed. in English), 52(6), 1722-1725 (2013-01-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service