Skip to Content
Merck
All Photos(1)

Key Documents

78891

Supelco

4-Hydroxy-4-methyl-2-pentanone

analytical standard

Synonym(s):

Diacetone alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$384.00

$384.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$384.00

About This Item

Linear Formula:
(CH3)2C(OH)CH2COCH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1740440
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$384.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

4 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.9 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.423 (lit.)

bp

166 °C (lit.)

density

0.931 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)CC(C)(C)O

InChI

1S/C6H12O2/c1-5(7)4-6(2,3)8/h8H,4H2,1-3H3

InChI key

SWXVUIWOUIDPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Hydroxy-4-methyl-2-pentanone may be used as an internal standard in the analysis of volatile odor-active compounds present in wine using solid phase extraction with gas chromatography (SPE-GC).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Building Blocks of the Chemical Industry (1989)
Determination of minor and trace volatile compounds in wine by solid-phase extraction and gas chromatography with mass spectrometric detection.
Lopez, et al.
Journal of Chromatography A, 966(1-2), 167-177 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service